siRNA Details
| Gene Symbol: Hhipl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019671.1 | - | 2 | 734 | ||
| BC034362.1 | - | 1 | 479 | 3154701 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hhipl2| Gene Symbol: Hhipl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019671.1 | - | 2 | 734 | ||
| BC034362.1 | - | 1 | 479 | 3154701 | ||