siRNA Details
| Gene Symbol: Kctd18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159864.1 | NP_001153336.1 | 3 | 406 | ||
| GenBank mRNA | AK032658.1 | - | 3 | 375 | ||
| AK053098.1 | - | 2 | 2358 | |||
| AK079700.1 | - | 3 | 402 | |||
| AK132699.1 | - | 3 | 375 | |||
| BC075692.1 | - | 3 | 460 | 30631161 | ||
| BY736102.1 | - | 3 | 378 | |||

Kctd18
Genomic Map