siRNA Details
| Gene Symbol: Dlst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030225.4 | NP_084501.1 | 4 | 242 | ||
| GenBank mRNA | AK019713.2 | - | 239 | |||
| AK054053.1 | - | 4 | 196 | |||
| AK149664.1 | - | 4 | 218 | |||
| AK158877.1 | - | 4 | 243 | |||
| AK168570.1 | - | 4 | 218 | |||
| AK169943.1 | - | 4 | 196 | |||
| AK202813.1 | - | 2 | 94 | |||
| BC006702.1 | - | 4 | 179 | 3583936 | ||
| CT010197.1 | - | 4 | 178 | |||

Dlst
Genomic Map