siRNA Details
| Gene Symbol: Gpr146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038703.2 | NP_001033792.1 | 2 | 1211 | ||
| NM_030258.4 | NP_084534.2 | 2 | 1136 | |||
| GenBank mRNA | AK046512.1 | - | 2 | 1159 | ||
| AK050034.1 | - | 2 | 1105 | |||
| AK087100.1 | - | 2 | 1105 | |||
| AK089232.1 | - | 2 | 1159 | |||
| AK137454.1 | - | 2 | 1213 | |||
| AK154478.1 | - | 2 | 1165 | |||
| AK154708.1 | - | 2 | 1157 | |||
| AK155212.1 | - | 2 | 1104 | |||
| AK155280.1 | - | 2 | 1225 | |||
| AK155876.1 | - | 2 | 1139 | |||
| BC003323.1 | - | 2 | 1107 | 3156060 | ||
More Information
Other Species Targeted
Set Membership: |
|

Gpr146
Genomic Map