siRNA Details
| Gene Symbol: Tmem50b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030018.3 | NP_084294.1 | 7 | 1005 | ||
| GenBank mRNA | AK020977.1 | - | 7 | 1010 | ||
| AK081455.1 | - | 1 | 24 | |||
| AK098154.1 | - | 7 | 969 | |||
| AK133966.1 | - | 7 | 1033 | |||
| AK150773.1 | - | 993 | ||||
| AK151796.1 | - | 7 | 1035 | |||
| AK152190.1 | - | 7 | 1021 | |||
| AK152767.1 | - | 7 | 1021 | |||
| AK155082.1 | - | 7 | 1058 | |||
| AK170684.1 | - | 7 | 1000 | |||
| AK172335.1 | - | 7 | 993 | |||
| AK183559.1 | - | 1 | 134 | |||
| BC004841.1 | - | 7 | 1008 | 2649264 | ||

Tmem50b
Genomic Map