siRNA Details
| Gene Symbol: Htra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030127.3 | NP_084403.2 | 9 | 2422 | ||
| GenBank mRNA | AK035194.1 | - | 9 | 2425 | ||
| AY037300.1 | - | 9 | 2210 | |||
| AY156509.1 | - | 9 | 2114 | |||
| BC056216.1 | - | 1 | 461 | |||

Htra3
Genomic Map