siRNA Details
| Gene Symbol: Zfp449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030139.4 | NP_084415.1 | 5 | 1844 | ||
| GenBank mRNA | AK036596.1 | - | 5 | 1845 | ||
| AK142888.1 | - | 5 | 1850 | |||
| BC082594.1 | - | 4 | 1734 | 30642886 | ||
| BC145936.1 | - | 4 | 1734 | 40131239 | ||
| BC145938.1 | - | 4 | 1734 | 40131241 | ||

Zfp449
Genomic Map