siRNA Details
| Gene Symbol: Naa35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030153.2 | NP_084429.2 | 11, 12 | 1014 | ||
| GenBank mRNA | AK021042.1 | - | 11, 12 | 905 | ||
| AK050805.1 | - | 1, 2 | 331 | |||
| AY102701.1 | - | 11, 12 | 936 | |||
| BC027201.1 | - | 11, 12 | 950 | |||
| BC056435.1 | - | 11, 12 | 923 | 5707801 | ||

Naa35
Genomic Map