siRNA Details
| Gene Symbol: Clip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271483.1 | NP_001258412.1 | 13 | 1749 | ||
| NM_001271484.1 | NP_001258413.1 | 13 | 1834 | |||
| NM_030179.3 | NP_084455.2 | 13 | 1737 | |||
| NM_175378.1 | NP_780587.1 | 12 | 1693 | |||
| GenBank mRNA | AK017914.1 | - | 10 | 1325 | ||
| AK019516.1 | - | 13 | 1750 | |||
| AK029962.1 | - | 13 | 1890 | |||
| AK075758.1 | - | 12 | 1684 | |||
| AK161301.1 | - | 13 | 1824 | |||
| AK163644.1 | - | 6 | 677 | |||
| BC011280.1 | - | 6 | 601 | |||
| BC035226.1 | - | 13 | 1737 | 4920788 | ||

Clip4
Genomic Map