siRNA Details
| Gene Symbol: Hspa13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030201.3 | NP_084477.1 | 5 | 1861 | ||
| GenBank mRNA | AK005396.1 | - | 1 | 678 | ||
| AK030553.1 | - | 6 | 1930 | |||
| AK034643.1 | - | 5 | 1841 | |||
| BC039701.1 | - | 4 | 1802 | |||
| BC085181.1 | - | 5 | 1822 | 30646642 | ||
| BC094508.1 | - | 6 | 1900 | |||

Hspa13
Genomic Map