siRNA Details
| Gene Symbol: Trim42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030219.2 | NP_084495.2 | 3 | 1336 | ||
| GenBank mRNA | AK019654.1 | - | 3 | 1337 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim42| Gene Symbol: Trim42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030219.2 | NP_084495.2 | 3 | 1336 | ||
| GenBank mRNA | AK019654.1 | - | 3 | 1337 | ||