siRNA Details
| Gene Symbol: Smim12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030252.2 | NP_084528.1 | 2 | 831 | ||
| GenBank mRNA | AK131844.1 | - | 1 | 309 | ||
| AK131915.1 | - | 2 | 796 | |||
| AK141221.1 | - | 2 | 765 | |||
| BC147067.1 | - | 2 | 793 | 9056070 | ||
| BC147068.1 | - | 2 | 793 | 9056074 | ||

Smim12
Genomic Map