siRNA Details
| Gene Symbol: Tusc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028182.1 | - | 1662 | |||
| AK081223.1 | - | 596 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tusc3| Gene Symbol: Tusc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028182.1 | - | 1662 | |||
| AK081223.1 | - | 596 | ||||