siRNA Details
| Gene Symbol: Rab27b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB067775.1 | - | 6 | 1253 | ||
| AF328893.1 | - | 6 | 1266 | |||
| AK049854.1 | - | 7 | 1150 | |||
| BC018443.1 | - | 6 | 1234 | 3601865 | ||
| BC032928.1 | - | 6 | 1186 | 5120275 | ||
| BC096496.1 | - | 7 | 1662 | 30738460 | ||

Rab27b
Genomic Map