siRNA Details
| Gene Symbol: BC004004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030561.3 | NP_085038.3 | 3 | 406 | ||
| GenBank mRNA | AK157037.1 | - | 3 | 223 | ||
| AK217020.1 | - | 2 | 196 | |||
| BC004004.1 | - | 3 | 243 | 3492768 | ||
| BC058575.1 | - | 3 | 215 | 6488611 | ||
| CD353313.1 | - | 3 | 358 | |||

BC004004
Genomic Map