siRNA Details
| Gene Symbol: Lrfn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001141921.1 | NP_001135393.1 | 4 | 721 | ||
| NM_030562.2 | NP_085039.2 | 4 | 682 | |||
| GenBank mRNA | AB243740.1 | - | 1 | 503 | ||
| AK090175.1 | - | 4 | 722 | |||
| AK144493.1 | - | 4 | 723 | |||
| BC004018.1 | - | 3 | 629 | 3494023 | ||
| DQ070870.1 | - | 1 | 503 | |||
| DQ314497.1 | - | 1 | 503 | |||

Lrfn1
Genomic Map