siRNA Details
| Gene Symbol: Trim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271725.1 | NP_001258654.1 | 5 | 887 | ||
| NM_001271726.1 | NP_001258655.1 | 5 | 923 | |||
| NM_001271727.1 | NP_001258656.1 | 5 | 1192 | |||
| NM_001271728.1 | NP_001258657.1 | 5 | 990 | |||
| NM_030706.3 | NP_109631.2 | 5 | 986 | |||
| GenBank mRNA | AB043550.1 | - | 5 | 970 | ||
| AF220017.1 | - | 5 | 835 | |||
| AK039741.1 | - | 5 | 984 | |||
| AK045410.1 | - | 5 | 983 | |||
| AK079415.1 | - | 5 | 1137 | |||
| AK141419.1 | - | 5 | 833 | |||
| AK147275.1 | - | 5 | 1100 | |||
| AK147395.1 | - | 5 | 981 | |||
| AK220343.1 | - | 5 | 1191 | |||
| BC058668.1 | - | 5 | 954 | |||
| BC058961.1 | - | 5 | 990 | 6826143 | ||

Trim2
Genomic Map