siRNA Details
| Gene Symbol: Vmn1r65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030738.2 | NP_109663.1 | 3 | 1388 | ||
| GenBank mRNA | AF324870.1 | - | 3 | 1389 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r65| Gene Symbol: Vmn1r65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030738.2 | NP_109663.1 | 3 | 1388 | ||
| GenBank mRNA | AF324870.1 | - | 3 | 1389 | ||