siRNA Details
| Gene Symbol: Sil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030749.2 | NP_109674.2 | 3 | 299 | ||
| GenBank mRNA | AJ297884.1 | - | 3 | 240 | ||
| AK170415.1 | - | 3 | 252 | |||
| BC016119.1 | - | 3 | 276 | 4489217 | ||
| BC016466.1 | - | 3 | 262 | 3582798 | ||
| CX219879.1 | - | 3 | 299 | |||

Sil1
Genomic Map