siRNA Details
| Gene Symbol: Lrba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077687.1 | NP_001071155.1 | 3 | 554 | ||
| NM_001077688.1 | NP_001071156.1 | 3 | 554 | |||
| NM_030695.2 | NP_109620.2 | 3 | 554 | |||
| GenBank mRNA | AF187731.1 | - | 3 | 576 | ||
| AF188506.1 | - | 3 | 576 | |||
| AF188507.1 | - | 3 | 576 | |||
| AK052148.1 | - | 3 | 741 | |||
| AK085208.1 | - | 3 | 543 | |||

Lrba
Genomic Map