siRNA Details
| Gene Symbol: Fcrls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF302046.1 | - | 3 | 244 | ||
| AY506556.1 | - | 3 | 215 | |||
| AY506557.1 | - | 3 | 215 | |||
| BC016551.1 | - | 3 | 218 | 3602803 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fcrls| Gene Symbol: Fcrls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF302046.1 | - | 3 | 244 | ||
| AY506556.1 | - | 3 | 215 | |||
| AY506557.1 | - | 3 | 215 | |||
| BC016551.1 | - | 3 | 218 | 3602803 | ||