siRNA Details
| Gene Symbol: Dtx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030714.2 | NP_109639.1 | 5 | 1223 | ||
| GenBank mRNA | AB015425.1 | - | 4 | 1205 | ||
| AK148444.1 | - | 5 | 1242 | |||
| AK180250.1 | - | 1 | 23 | |||
| AK180771.1 | - | 1 | 23 | |||
| AK181396.1 | - | 1 | 23 | |||
| AK183648.1 | - | 1 | 23 | |||
| AK189235.1 | - | 1 | 23 | |||
| AK195108.1 | - | 1 | 23 | |||
| AK205640.1 | - | 1 | 41 | |||
| AK207039.1 | - | 1 | 23 | |||
| AK218545.1 | - | 1 | 23 | |||
| AK220046.1 | - | 1 | 23 | |||
| BC099687.1 | - | 5 | 1223 | 6311735 | ||
| BC138304.1 | - | 4 | 1102 | 8861323 | ||
| BC157899.1 | - | 4 | 1104 | 9088091 | ||

Dtx3
Genomic Map