siRNA Details
| Gene Symbol: Lactb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030717.1 | NP_109642.1 | 3, 4 | 701 | ||
| GenBank mRNA | AF317900.1 | - | 3, 4 | 701 | ||
| AK078550.1 | - | 3, 4 | 642 | |||
| AK087297.1 | - | 3, 4 | 670 | |||
| AK149410.1 | - | 3, 4 | 644 | |||
| BC016583.1 | - | 3, 4 | 628 | |||
| BC021604.1 | - | 3, 4 | 627 | |||
| BC046293.1 | - | 3, 4 | 628 | 6484767 | ||
| BC147809.1 | - | 3, 4 | 646 | 9087526 | ||

Lactb
Genomic Map