siRNA Details
| Gene Symbol: Abo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030718.5 | NP_109643.3 | 4, 5 | 281 | ||
| GenBank mRNA | AB041039.1 | - | 3, 4 | 169 | ||
| AK033786.1 | - | 4, 5 | 202 | |||
| AK035261.1 | - | 4, 5 | 266 | |||
| BC116759.1 | - | 4, 5 | 230 | 40126078 | ||
| BC116761.1 | - | 4, 5 | 230 | 40126080 | ||

Abo
Genomic Map