siRNA Details
| Gene Symbol: BC004004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030561.3 | NP_085038.3 | 6 | 1007 | ||
| GenBank mRNA | AK030596.1 | - | 4 | 577 | ||
| AK180627.1 | - | 2 | 142 | |||
| AK180865.1 | - | 228 | ||||
| AK191458.1 | - | 2 | 142 | |||
| AK198839.1 | - | 2 | 142 | |||
| BC004004.1 | - | 6 | 844 | 3492768 | ||
| BC058575.1 | - | 6 | 816 | 6488611 | ||

BC004004
Genomic Map