siRNA Details
| Gene Symbol: Slc14a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110274.1 | NP_001103744.1 | 2 | 249 | ||
| NM_030683.3 | NP_109608.1 | 6 | 875 | |||
| NM_207651.3 | NP_997534.2 | 6 | 875 | |||
| GenBank mRNA | AF258601.2 | - | 2 | 249 | ||
| AF258602.1 | - | 6 | 776 | |||
| AF366052.2 | - | 6 | 867 | |||
| BC150680.1 | - | 6 | 783 | 9087591 | ||

Slc14a2
Genomic Map