siRNA Details
| Gene Symbol: Zfp202 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030713.2 | NP_109638.2 | 7 | 1713 | ||
| GenBank mRNA | AF292648.1 | - | 7 | 1703 | ||
| AF292649.1 | - | 7 | 1699 | |||
| AK170181.1 | - | 6 | 1585 | |||
| AK172368.1 | - | 7 | 1713 | |||
| BC049772.1 | - | 7 | 1641 | 6334069 | ||

Zfp202
Genomic Map