siRNA Details
| Gene Symbol: Lactb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030717.1 | NP_109642.1 | 4 | 981 | ||
| GenBank mRNA | AF317900.1 | - | 4 | 981 | ||
| AK078550.1 | - | 4 | 922 | |||
| AK213700.1 | - | 1 | 51 | |||
| BC046293.1 | - | 4 | 908 | 6484767 | ||
| BC147809.1 | - | 4 | 926 | 9087526 | ||

Lactb
Genomic Map