siRNA Details
| Gene Symbol: Tbl1xr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030732.3 | NP_109657.2 | 15 | 1873 | ||
| GenBank mRNA | AF268195.1 | - | 15 | 1654 | ||
| AK029595.1 | - | 15 | 1672 | |||
| AK033347.1 | - | 15 | 1743 | |||
| AK036064.1 | - | 16 | 1984 | |||
| AK136255.1 | - | 15 | 1695 | |||
| BC018512.1 | - | 5 | 519 | |||

Tbl1xr1
Genomic Map