siRNA Details
| Gene Symbol: Sgpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030750.3 | NP_109675.1 | 3 | 2872 | ||
| GenBank mRNA | AK154116.1 | - | 4 | 2514 | ||
| BC026814.1 | - | 3 | 2808 | |||
| BC037592.1 | - | 3 | 2825 | 5004335 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sgpp1| Gene Symbol: Sgpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030750.3 | NP_109675.1 | 3 | 2872 | ||
| GenBank mRNA | AK154116.1 | - | 4 | 2514 | ||
| BC026814.1 | - | 3 | 2808 | |||
| BC037592.1 | - | 3 | 2825 | 5004335 | ||