siRNA Details
| Gene Symbol: Ift122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167763.1 | NP_001161235.1 | 2, 3 | 243 | ||
| NM_031177.4 | NP_112454.2 | 2, 3 | 243 | |||
| GenBank mRNA | AK031308.1 | - | 2, 3 | 131 | ||
| AK044456.1 | - | 2, 3 | 243 | |||
| AK131115.1 | - | 2, 3 | 170 | |||
| AK167330.1 | - | 2, 3 | 198 | |||
| AK167412.1 | - | 2, 3 | 198 | |||
| AK199875.1 | - | 1, 2 | 72 | |||
| BC060621.1 | - | 2, 3 | 191 | |||
| BC066083.1 | - | 2, 3 | 162 | 30355922 | ||
| BC067415.1 | - | 2, 3 | 159 | 30063124 | ||

Ift122
Genomic Map