siRNA Details
| Gene Symbol: Plekha3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031256.3 | NP_112546.1 | 2 | 403 | ||
| GenBank mRNA | AF286163.1 | - | 2 | 192 | ||
| BC012283.1 | - | 2 | 143 | |||
| BC031110.1 | - | 3 | 211 | 4981486 | ||
| BY730332.1 | - | 2 | 403 | |||

Plekha3
Genomic Map