siRNA Details
| Gene Symbol: Pik3ap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031376.3 | NP_113553.1 | 2 | 193 | ||
| GenBank mRNA | AF293806.1 | - | 2 | 193 | ||
| AK087295.1 | - | 2 | 220 | |||
| AK089785.1 | - | 2 | 226 | |||
| AK154326.1 | - | 2 | 221 | |||
| AK170130.1 | - | 2 | 226 | |||
| AK170925.1 | - | 2 | 226 | |||
| AK171021.1 | - | 2 | 221 | |||
| BC113141.1 | - | 2 | 152 | 40047744 | ||
| BY206955.1 | - | 2 | 241 | |||

Pik3ap1
Genomic Map