siRNA Details
| Gene Symbol: Usp26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031388.2 | NP_113565.2 | 8 | 1072 | ||
| GenBank mRNA | AF285570.1 | - | 5 | 668 | ||
| AK082829.1 | - | 8 | 1000 | |||
| AK133417.1 | - | 8 | 1076 | |||
| BC125454.1 | - | 3 | 571 | 40129969 | ||
| BC132154.1 | - | 3 | 571 | 40130431 | ||

Usp26
Genomic Map