siRNA Details
| Gene Symbol: Sytl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040085.2 | NP_001035174.1 | 2, 3 | 852 | ||
| NM_001289583.1 | NP_001276512.1 | 2, 3 | 852 | |||
| NM_001289584.1 | NP_001276513.1 | 2, 3 | 852 | |||
| NM_031394.3 | NP_113571.2 | 2, 3 | 852 | |||
| GenBank mRNA | AB057754.1 | - | 1, 2 | 191 | ||
| AB057760.1 | - | 1, 2 | 191 | |||
| AB057761.1 | - | 1, 2 | 191 | |||
| AB057762.1 | - | 1, 2 | 191 | |||
| BC052915.1 | - | 1, 2 | 196 | |||
| BC064769.1 | - | 1, 2 | 184 | |||
| BC138714.1 | - | 1, 2 | 162 | 8861736 | ||
| BC145398.1 | - | 1, 2 | 162 | 9053956 | ||

Sytl2
Genomic Map