siRNA Details
| Gene Symbol: Gigyf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031408.2 | NP_113596.2 | 3 | 254 | ||
| GenBank mRNA | BC052875.1 | - | 3 | 232 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gigyf1| Gene Symbol: Gigyf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031408.2 | NP_113596.2 | 3 | 254 | ||
| GenBank mRNA | BC052875.1 | - | 3 | 232 | ||