siRNA Details
| Gene Symbol: Tdrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002238.2 | NP_001002238.1 | 9 | 1107 | ||
| NM_001002240.2 | NP_001002240.1 | 9 | 1080 | |||
| NM_001002241.2 | NP_001002241.1 | 8 | 1380 | |||
| NM_031387.3 | NP_113564.2 | 9 | 1240 | |||
| GenBank mRNA | AB067571.1 | - | 8 | 1061 | ||
| AB183526.1 | - | 9 | 1107 | |||
| AB183527.1 | - | 9 | 1080 | |||
| AB183528.1 | - | 8 | 1380 | |||
| AB183529.1 | - | 8 | 944 | |||
| AF285591.1 | - | 5 | 434 | |||
| AK029806.1 | - | 9 | 1241 | |||
| BC129954.1 | - | 4 | 366 | 40081321 | ||
| BC129955.1 | - | 4 | 366 | 40081323 | ||

Tdrd1
Genomic Map