siRNA Details
| Gene Symbol: Sytl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB050741.1 | - | 5, 6 | 589 | ||
| AK154176.1 | - | 5, 6 | 605 | |||
| BC005623.1 | - | 5, 6 | 610 | 3155535 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sytl1| Gene Symbol: Sytl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB050741.1 | - | 5, 6 | 589 | ||
| AK154176.1 | - | 5, 6 | 605 | |||
| BC005623.1 | - | 5, 6 | 610 | 3155535 | ||