siRNA Details
| Gene Symbol: Bicc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031397.2 | NP_113574.1 | 3 | 278 | ||
| GenBank mRNA | AF319464.1 | - | 3 | 279 | ||
| AK076154.1 | - | 3 | 331 | |||
| AK089066.1 | - | 3 | 109 | |||
| BC111523.2 | - | 3 | 279 | 40048106 | ||

Bicc1
Genomic Map