siRNA Details
| Gene Symbol: Pde4dip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039376.2 | NP_001034465.2 | 47 | 8183 | ||
| NM_001289701.1 | NP_001276630.1 | 47 | 8234 | |||
| NM_001289702.1 | NP_001276631.1 | 46 | 8081 | |||
| GenBank mRNA | AJ290946.1 | - | 1 | 557 | ||
| AK040750.1 | - | 8 | 1821 | |||
| BC022211.1 | - | 1 | 665 | |||
| BC046620.1 | - | 1 | 740 | |||
| BC141185.1 | - | 47 | 8234 | 9055797 | ||
| BC145504.1 | - | 46 | 8081 | 9054067 | ||

Pde4dip
Genomic Map