siRNA Details
| Gene Symbol: Akap12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031185.3 | NP_112462.1 | 4 | 5759 | ||
| GenBank mRNA | AB020886.1 | - | 4 | 5708 | ||
| AB025278.1 | - | 4 | 5722 | |||
| AB051563.1 | - | 4 | 5710 | |||
| AB070852.1 | - | 3 | 5444 | |||
| AB070853.1 | - | 3 | 5430 | |||
| AF326228.1 | - | 4 | 5708 | |||
| AF326230.1 | - | 3 | 5413 | |||
| BC042461.1 | - | 2 | 5155 | |||
| BC043939.1 | - | 2 | 5155 | |||
| BC071185.1 | - | 4 | 5707 | |||

Akap12
Genomic Map