siRNA Details
Gene Symbol: Tex13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031381.2 | NP_113558.1 | 4 | 1818 | ||
GenBank mRNA | AF285576.1 | - | 4 | 1818 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tex13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031381.2 | NP_113558.1 | 4 | 1818 | ||
GenBank mRNA | AF285576.1 | - | 4 | 1818 |