siRNA Details
| Gene Symbol: Cnnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031396.2 | NP_113573.2 | 11 | 3228 | ||
| GenBank mRNA | AF202994.1 | - | 11 | 2454 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnnm1| Gene Symbol: Cnnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031396.2 | NP_113573.2 | 11 | 3228 | ||
| GenBank mRNA | AF202994.1 | - | 11 | 2454 | ||