siRNA Details
| Gene Symbol: STAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000349.2 | NP_000340.2 | 7 | 1336 | ||
| XM_006716392.2 | XP_006716455.1 | 6 | 1425 | |||
| GenBank mRNA | AF035277.1 | - | 7 | 1184 | ||
| AK090513.1 | - | 4 | 3000 | |||
| AY532319.1 | - | 1 | 302 | |||
| BC010550.1 | - | 7 | 1137 | 4153601 | ||
| BX647744.1 | - | 2 | 2157 | |||
| S79669.1 | - | 7 | 1223 | |||
| U17280.1 | - | 7 | 1197 | |||

STAR
Genomic Map