siRNA Details
| Gene Symbol: Zfp423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033327.2 | NP_201584.2 | 4 | 856 | ||
| GenBank mRNA | AF188609.1 | - | 2 | 225 | ||
| AK122365.1 | - | 4 | 822 | |||
| AK133497.1 | - | 2 | 284 | |||
| AY147407.1 | - | 3 | 461 | |||
| AY256893.1 | - | 4 | 755 | |||
| BC059234.1 | - | 2 | 667 | |||
| BC079586.1 | - | 4 | 542 | |||
| BC139028.1 | - | 4 | 602 | 8862050 | ||
| BC139030.1 | - | 4 | 602 | 8862052 | ||

Zfp423
Genomic Map