siRNA Details
| Gene Symbol: Sec16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159986.1 | NP_001153458.1 | 2 | 187 | ||
| NM_033354.3 | NP_203505.3 | 4 | 434 | |||
| GenBank mRNA | AB063356.1 | - | 2 | 187 | ||
| AK132144.1 | - | 2 | 211 | |||
| AK173302.1 | - | 4 | 415 | |||
| BC059194.1 | - | 3 | 252 | 4222915 | ||

Sec16b
Genomic Map