siRNA Details
| Gene Symbol: Cnnm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102471.1 | NP_001095941.1 | 1 | 889 | ||
| NM_033569.3 | NP_291047.2 | 1 | 889 | |||
| GenBank mRNA | AF216961.1 | - | 1 | 201 | ||
| AK032006.1 | - | 1 | 849 | |||
| AK044400.1 | - | 1 | 893 | |||
| AK053711.1 | - | 1 | 907 | |||
| AK159616.1 | - | 1 | 895 | |||
| BC012956.1 | - | 1 | 885 | |||
| BC027387.1 | - | 1 | 686 | |||
| BC125033.1 | - | 1 | 201 | |||
| GQ225745.1 | - | 1 | 188 | |||

Cnnm2
Genomic Map