siRNA Details
| Gene Symbol: Tmem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033759.2 | NP_001028931.1 | 11, 12 | 2645 | ||
| NM_031997.4 | NP_114386.3 | 12, 13 | 2727 | |||
| GenBank mRNA | AK129352.1 | - | 6, 7 | 946 | ||
| BC019745.1 | - | 3, 4 | 368 | 3675225 | ||
| BC076570.1 | - | 11, 12 | 2251 | 6830791 | ||
| BC089353.1 | - | 11, 12 | 2627 | 6410437 | ||
| BU058172.1 | - | 2, 3 | 159 | |||
| CA752446.1 | - | 4, 5 | 585 | |||

Tmem2
Genomic Map