siRNA Details
| Gene Symbol: Trps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032000.2 | NP_114389.2 | 2 | 527 | ||
| GenBank mRNA | AF346836.1 | - | 3 | 452 | ||
| AK142355.1 | - | 2 | 197 | |||
| BC037058.1 | - | 2 | 205 | 4951294 | ||
| BC049857.1 | - | 3 | 231 | 4460789 | ||
| BC099962.1 | - | 2 | 492 | 30091008 | ||

Trps1
Genomic Map