siRNA Details
| Gene Symbol: M1ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033079.2 | NP_149070.2 | 3 | 347 | ||
| GenBank mRNA | AF084364.1 | - | 2 | 316 | ||
| AK131954.1 | - | 3 | 347 | |||
| BC119803.1 | - | 2 | 274 | 40107692 | ||
| BC122521.1 | - | 2 | 279 | 40107688 | ||

M1ap
Genomic Map